Natriumdiethylditiokarbamat trihydrat ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: (Diethyldithiocarbamato-S,S')sodium trihydrate, Diethyldithiocarbamic acid sodium salt trihydrate, N,N-Diethyldithiocarbamic acid sodium salt trihydrate
A15898.22 A15898.36
A15898.22EA 1375 SEK
A15898.22 A15898.36
Natriumdiethylditiokarbamat trihydrat ≥98%
Natriumdiethylditiokarbamat trihydrat
Purity: 98% CAS No.: 20624-25-3 Molecular Formula: C5H10NNaS2.3H2O Formula Weight: 225.31 (171.28anhy) Melting Point: 94-102° Beilstein Reference: 3920507 Merck Reference: 14,3378 MDL No.: MFCD00150617
Formel: (C₂H₅)₂NCSSNa·3H₂O
MW: 225.31 g/mol
Smältpunkt: 94…102 °C
Densitet: 1.1 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00150617
CAS nummer: 20624-25-3
EINECS: 205-710-6
UN: 3077
ADR: 9 III
Merck Index: 12,03443