Tetrapropylammonium hydroxide 1 M i vattenlösning
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: IPC-TPA-OH
L14328.14 L14328.22 L14328.36
L14328.14EA 680 SEK
L14328.14 L14328.22 L14328.36
Tetrapropylammonium hydroxide 1 M i vattenlösning
Tetrapropylammonium hydroxide
CAS No.: 4499-86-9 Molecular Formula: (CH3CH2CH2)4NOH Formula Weight: 203.37 Beilstein Reference: 4207983 UN No.: 3267 MDL No.: MFCD00009360
Formel: C₁₂H₂₉NO
MW: 203.37 g/mol
Densitet: 1.00
Storage Temperature: Omgivande
MDL Number: MFCD00009360
CAS nummer: 4499-86-9
EINECS: 224-800-6
UN: 3267
ADR: 8 II