DL-Lysine monohydrochloride ≥99%
Leverantör: Thermo Scientific
Synonyms: (±)-2,6-Diaminocaproic acid monohydrochloride, H-DL-Lys-OH · HCl, H-DL-Lys-OH.HCl, (±)-2,6-Diaminohexanoic acid monohydrochloride, (±)-Lysine monohydrochloride
A11066.14 A11066.22
A11066.14EA 835 SEK
A11066.14 A11066.22
DL-Lysine monohydrochloride ≥99%
DL-Lysine monohydrochloride
Purity: 99% CAS No.: 70-53-1 Molecular Formula: C6H14N2O2.HCl Formula Weight: 182.65 Melting Point: 267° dec. Beilstein Reference: 4711993
Formel: C₆H₁₅ClN₂O₂
MW: 182,65 g/mol
Smältpunkt: 267 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00064563
CAS nummer: 70-53-1
EINECS: 200-739-0