Tetrapropylammonium hydroxide 25% i vatten
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: IPC-TPA-OH
420730250 420731000 420735000
ACRO420730250EA 635 SEK
ACRO420730250 ACRO420731000 ACRO420735000
Tetrapropylammonium hydroxide 25% i vatten
Tetrapropylammonium hydroxide
Formel: C₁₂H₂₉NO
MW: 203.37 g/mol
Densitet: 1 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00009360
CAS nummer: 4499-86-9
EINECS: 224-800-6
UN: 3267
ADR: 8 II
Order Now
Specifikation testresultat
Appearance Clear colorless Solution
Titration with HClO4 24 to 26 %
Melting point 114 to 118 °C
HPLC ≥96.0 %