Metyl(±)-laktat 99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Methyl Lactate, Methyl DL-lactate, Lactic acid methyll ester, Methyl (±)-2-hydroxypropionate
414480250 414480050
ACRO414480250EA 3000 SEK
ACRO414480250 ACRO414480050
Metyl(±)-laktat 99%
Metyl(±)-laktat
Formel: C₄H₈O₃
Kokpunkt: 35 °C (6 mmHg)
Densitet: 1,09 g/cm³
Flampunkt: 49 °C
Storage Temperature: Omgivande
MDL Number: MFCD00066367
CAS nummer: 547-64-8
EINECS: 208-930-0
UN: 3272
ADR: 3,III
Merck Index: 12,06167
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4120 to 1.4150 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation -0.5° to +0.5° (20°C, 589 nm) (neat)