Pentacarbonyliron
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Iron pentacarbonyl, Iron(0) pentacarbonyl
412600250 412601000
ACRO412600250EA 765 SEK
ACRO412600250 ACRO412601000
Pentacarbonyliron
Pentacarbonyliron
Formel: C₅FeO₅
MW: 195.9 g/mol
Kokpunkt: 103
Smältpunkt: –20 °C
Densitet: 1.49 g/cm³
Flampunkt: –15 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00011001
CAS nummer: 13463-40-6
EINECS: 236-670-8
UN: 1994
ADR: 6.1 I
Merck Index: 12,05111
Order Now
Specifikation testresultat
Appearance Yellow-orange to brown Liquid (May contain particulate matter)
Infrared spectrum Conforms
Additional info Material may have to be filtered through glass wool before use. Keep residue moist, may be pyrophoric if allowed to dry.
GC ≥99.5 %
Color scale ≤50 APHA