Metylorange 0,04% aqueous solution, TECHNICAL Indicator_pH
Leverantör: VWR Chemicals
Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt, Methyl orange sodium salt, Helianthine
31720.292EA 549 SEK
31720.292 31720.230
Metylorange 0,04% aqueous solution, TECHNICAL Indicator_pH
Metylorange
  • Transition range: pH 3,1-4,4
Formel: C₁₄H₁₄N₃NaO₃S
MW: 327.34 g/mol
Kokpunkt: 90
Flampunkt: 38
Storage Temperature: Omgivande
MDL Number: MFCD00007502
CAS nummer: 547-58-0
EINECS: 208-925-3
Merck Index: 13,06127
Order Now
Specifikation testresultat
Identification Passes test