Potassium dichromate 0.04 mol/l (0.24 N) with mercury (II) sulphate 80 g/l in sulphuric acid solution 0.04 mol/l (0.24 N) Mercury(II)-sulfate sulfuric acid_80g/l, AVS TITRINORM Volumetric, COD_determination_water
Leverantör: VWR Chemicals
Fara
30891.295EA 492 SEK
30891.295 30891.420
Potassium dichromate 0.04 mol/l (0.24 N) with mercury (II) sulphate 80 g/l in sulphuric acid solution 0.04 mol/l (0.24 N) Mercury(II)-sulfate sulfuric acid_80g/l, AVS TITRINORM Volumetric, COD_determination_water
Kaliumdikromat
According to NFT 90-101 standard
Formel: K₂Cr₂O₇
MW: 294.18 g/mol
Kokpunkt: >100 °C (1013 hPa)
Smältpunkt: 398
Densitet: 1.19 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011367
CAS nummer: 7778-50-9
EINECS: 231-906-6
UN: 3289
ADR: 6.1 (8) II
Merck Index: 12,07790
Order Now
Specifikation testresultat
Titer (20°C; real value 0.2 % accuracy) 0.03992 - 0.04008 mol/l
NIST traceable Confirmed