Sand Fontainebleau, TECHNICAL
Leverantör: VWR Chemicals
27460.295EA 360 SEK
27460.295 27460.364 27460.460
Sand Fontainebleau, TECHNICAL
Sand Fontainebleau
Formel: SiO₂
MW: 60,08 g/mol
Kokpunkt: 2230 °C (1013 hPa)
Storage Temperature: Omgivande
CAS nummer: 7631-86-9
EINECS: 238-878-4
Order Now
Specifikation testresultat
Identification Passes test