, vattenfri, LiChropur™ för HPLC, Supelco®
Leverantör: Merck
Synonyms: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate
M-Clarity™ quality level = MQ100
5.43839.0100 5.43839.0250
5.43839.0100EA 1775 SEK
5.43839.0100 5.43839.0250
, vattenfri, LiChropur™ för HPLC, Supelco®
Dikaliumhydrogenfosfat
Formel: K₂HPO₄
MW: 174.18 g/mol
Smältpunkt: 340 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011383
CAS nummer: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM