O-Methylhydroxylamine hydrochloride 97.5-102.5%, LiChropur™ for GC derivatisation, Supelco®
Leverantör: Merck
Fara
Synonyms: Methylhydroxylamine hydrochloride, O-Methylhydroxylammonium chloride, Methoxyamine hydrochloride, Methoxylamine hydrochloride
89803-1G 89803-5G
89803-1GEA 425 SEK
89803-1G 89803-5G
O-Methylhydroxylamine hydrochloride 97.5-102.5%, LiChropur™ for GC derivatisation, Supelco®
O-Methylhydroxylamine hydrochloride
Formel: CH₆ClNO
MW: 83.515 g/mol
Smältpunkt: 151...154 °C (lit.)
MDL Number: MFCD00012951
CAS nummer: 593-56-6
UN: 3261
ADR: 8 II
Order Now
Specifikation testresultat
Appearance (Color) White
Appearance (Form) Powder or Crystals
Argentometric Titration 97.5 - 102.5 %
Proton NMR Spectrum Conforms to Structure