Papanicolaou's solution 1b Hematoxylin solution S staining_nuclear, cytology, Sigma-Aldrich®
Leverantör: Merck
|
Varning
|
1.09254.0500
1.09254.2500
1.09254.0500EA
545
SEK
1.09254.0500
1.09254.2500
Papanicolaou's solution 1b Hematoxylin solution S staining_nuclear, cytology, Sigma-Aldrich®
Papanicolaou's solution 1b Hematoxylin solution S
Nuclear staining in cytology
The cytological staining technique developed by Papanicolaou is still the standard method in cancer and hormone cycle diagnosis. The modified hematoxylin solution S (Papanicolaou’s solution 1b) provides good nuclear staining in 2-3 minutes. Nuclei are stained blue, dark violet to black. A 500 ml bottle is sufficient for 1500 up to 2500 slides. It is registered as IVD and CE certified product and can be used for clinical diagnostics.
![]() |
Specifikation testresultat
Suitability for microscopy (Vaginal smear) | passes test |
Nuclei | blue to dark violet |
Cyanophilic cytoplasm | blue-green |
Eosinophilic cytoplasm | pink |