N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane Silyl-991 silylation reagent, for GC derivatisation
Leverantör: MACHEREY-NAGEL
Varning
Synonyms: BSTFA+TMCS (99:1)
701490.1100 701490.150 701490.201
MANA701490.1100EA 17225 SEK
MANA701490.1100 MANA701490.150 MANA701490.201
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane Silyl-991 silylation reagent, for GC derivatisation
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane
Silyl-991 is recommended for silylation of fatty acid amides, hindered hydroxyls and other compounds which are difficult to silylate (e.g., secondary alcoholes and amines).
  • Silylation can be applied on many compounds
  • Silylation reagents are easily prepared
  • In Silyl-991 mixture the strong TMS donor BSTFA is catalysed with 1% TMCS
Silyl-991 silylating reagent is a mixture of BSTFA + TMCS (99:1) without pyridene.
Formel: C₈H₁₈F₃NOSi₂
MW: 257,40 g/mol
Kokpunkt: 40 °C (12 mm Hg)
Densitet: 0,97 g/cm³
Flampunkt: 7 °C
MDL Number: MFCD00008269
CAS nummer: 25561-30-2
UN: 3316
ADR: 9,II