N, O-bis (trimetylsilyl)acetamid ≥98.5%, LiChropur™ for GC derivatisation, Supelco®
Leverantör: Merck
Synonyms: BSA, Silyl Derivatisation, Trimethylsilyl N-(trimethylsilyl)acetimidate, Trimethylsilyl N-trimethylsilylacetamidate
15269-25ML 15269-5ML 15269-10X1ML
15269-25MLEA 2420 SEK
15269-25ML 15269-5ML 15269-10X1ML
N, O-bis (trimetylsilyl)acetamid ≥98.5%, LiChropur™ for GC derivatisation, Supelco®
N, O-bis (trimetylsilyl)acetamid
Formel: C₈H₂₁NOSi₂
MW: 203.43 g/mol
Kokpunkt: 162
Smältpunkt: 24
Flampunkt: 40
MDL Number: MFCD00008270
CAS nummer: 10416-59-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0a40f8f42759b8bc90a1c4436b6b05137a0607e&vid=b0a40f8f42759b8bc90a1c4436b6b05137a0607e&prid=web&clid=SAPDAM