Safranine 0 (3,7-diamino-2,8-dimetyl-5-fenylfenaziniumklorid) lösning for Gram staining, Sigma-Aldrich®
Leverantör: Merck
Varning
Synonyms: CI 50240
M-Clarity™ quality level = MQ300
1.09217.0500 1.09217.2500
1.09217.0500EA 362 SEK
1.09217.0500 1.09217.2500
Safranine 0 (3,7-diamino-2,8-dimetyl-5-fenylfenaziniumklorid) lösning for Gram staining, Sigma-Aldrich®
Safranine 0 (3,7-diamino-2,8-dimetyl-5-fenylfenaziniumklorid)
Counterstain for Gram staining
The Gram staining allows a fast differentiation of bacteria in Gram-positive and Gram-negative. The mureine structure of the bacteria walls is the basis of the color affinity. Bacteria will be stained with Gram's crystal violet solution - an aniline dye - in the first step. After the treatment with iod solution (Lugol’s solution), a dye-iod complex will form. During the decolorizing step, this complex stays in the multilayer mureine structures of the Gram-positive bacteria and they will appear blue. Gram-negative bacteria have a monolayer mureine structure only, the dye-iod complex does not stay bound to the cellwall, they will be decolorized . Gram-negative bacteria will be counterstained by Gram's safranin solution and appear orange. A bottle with 500 ml Gram's safranin solution is sufficient for staining up to 250 slides, it is also available as part of the Gram-color stain set. It is an IVD product and CE registered.
Formel: C₂₀H₁₉ClN₄
MW: 350,85 g/mol
Densitet: 0,98 g/cm3 (20 °C)
Flampunkt: 49 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011759
CAS nummer: 477-73-6
IVDLogo
Order Now
Specifikation testresultat
Suitability for microscopy passes test
gram-positve microorganisms blue-violet
gram-negative microorganisms pink to red