N-Metyl-2-pyrrolidon (NMP) ≥99.5%, LiChrosolv® for liquid chromatography, Supelco®
Leverantör: Merck
Fara
Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone
M-Clarity™ quality level = MQ100
5.43899.1000 5.43899.2500
5.43899.1000EA 1670 SEK
5.43899.1000 5.43899.2500
N-Metyl-2-pyrrolidon (NMP) ≥99.5%, LiChrosolv® for liquid chromatography, Supelco®
N-Metyl-2-pyrrolidon (NMP)
Formel: C₅H₉NO
MW: 99.13 g/mol
Kokpunkt: 202 °C (1013 hPa)
Smältpunkt: –24 °C
Flampunkt: 91 °C
MDL Number: MFCD00003193
CAS nummer: 872-50-4
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Purity (GC) ≥ 99.5 %
Water ≤ 0.05 %
Transmission (at 300 nm) ≥ 50 %
Transmission (at 325 nm) ≥ 90 %
Transmission (at 350 nm) ≥ 97 %
Transmission (at 400 nm) ≥ 99 %