Sisomicin ≥98%
Leverantör: Cayman Chemical
|
Fara
|
Synonyms:
Rickamicin, Salvamina, SCH 13475, Antibiotic 6640
Sisomicin is a broad-spectrum aminoglycoside antibiotic originally isolated from M. inyoensis.
It is active against Gram-positive and Gram-negative bacteria including B. subtilis, S. aureus, E. coli, and P. aeruginosa among others (MICs = 0.01-0.75 µg/ml) but is less active against P. multocida and V. coli (MICs = 7.5 and 2.4 µg/ml, respectively). Sisomicin is protective against Gram-positive and Gram-negative bacterial infections in mice with 50% protective dose (PD50s) values ranging from 0.5 to 6 mg/kg.
Formel:
C₁₉H₃₇N₅O₇ MW: 447,50 g/mol Storage Temperature: Frys |
MDL Number:
MFCD00865123 CAS nummer: 32385-11-8 EINECS: 251-018-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...