Alfacalcidol ≥95%, crystalline solid
Leverantör: Cayman Chemical
Fara
Synonyms: (1R,3S,Z)-5-((E)-2-((1R,3aS,7aR)-7a-Methyl-1-((R)-6-methylheptan-2-yl)hexahydro-1H-inden-4(2H)-ylidene)ethylidene)-4-methylenecyclohexane-1,3-diol, 1alpha-Hydroxyvitamin D3, 1alpha-Hydroxycholecalciferol
12082-1 12082-5 12082-10
CAYM12082-1EA 1100 SEK
CAYM12082-1 CAYM12082-5 CAYM12082-10
Alfacalcidol ≥95%, crystalline solid
Alfacalcidol
Formel: C₂₇H₄₄O₂
MW: 400,65 g/mol
Storage Temperature: Frys
MDL Number: MFCD00868328
CAS nummer: 41294-56-8
UN: 2811
ADR: 6.1,II
Order Now
Specifikation testresultat
Purity ≥95%
λmax 213, 265 nm