Sand, quartz 50-70 mesh, Fluka™
Leverantör: Honeywell Chemicals
Fara
274739-1KG 274739-5KG 274739-500G
274739-1KGEA 1125 SEK
274739-1KG 274739-5KG 274739-500G
Sand, quartz 50-70 mesh, Fluka™
Sand, quartz
Formel: SiO₂
MW: 60,08 g/mol
Kokpunkt: 2,210 °C (1.013 hPa)
Smältpunkt: 1,710 °C
Densitet: 2,650 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011232
CAS nummer: 14808-60-7
EINECS: 238-878-4
Order Now
Specifikation testresultat
Particle Size 50-70 mesh
Appearance complying
Granulation 0.21-0.3 mm Min. 85 %