Tymolblått indicator S. No.: 879, Fluka™
Leverantör: Honeywell Chemicals
Synonyms: Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
32728-5G 32728-25G
32728-5GEA 705 SEK
32728-5G 32728-25G
Tymolblått indicator S. No.: 879, Fluka™
Tymolblått
Formel: C₂₇H₃₀O₅S
MW: 466.6 g/mol
Smältpunkt: 221 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005869
CAS nummer: 76-61-9
EINECS: 200-973-3
Merck Index: 12,09541
Order Now
Specifikation testresultat
Identity (IR) complying
Change range (Ph1,2 - 2,8) complying
Change range (PH₈,0 - 9,6) complying
Indicator solution complying
Extinction maximum at 589 - 599 nm
Extinction maximum of 700 - 900