Natriumtiosulfat concentrate for dilution to 1L, 0.1 M Na2S2O3 (0.1N) volumetrisk lösning, Fluka™
Leverantör: Honeywell Chemicals
Synonyms: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
38200-1EA 38200-6X1EA
38200-1EAEA 370 SEK
38200-1EA 38200-6X1EA
Natriumtiosulfat concentrate for dilution to 1L, 0.1 M Na2S2O3 (0.1N) volumetrisk lösning, Fluka™
Natriumtiosulfat
Concentrate for 1L standard solution
Formel: Na₂S₂O₃
MW: 158.11 g/mol
Kokpunkt: 100 °C
Smältpunkt: 48
Flampunkt: Does not flash
Storage Temperature: Omgivande
MDL Number: MFCD00003499
CAS nummer: 7772-98-7
EINECS: 231-791-2
Merck Index: 12,08844
Order Now
Specifikation testresultat
Factor 0.998 - 1.002