Salpetersyra ≥69,0%, TraceSELECT™ för spåranalys, Fluka™
Leverantör: Honeywell Chemicals
Fara
Synonyms: Nitric acid solution
84385-500ML 84385-6X500ML 84385-1L 84385-2.5L
84385-500MLEA 1375 SEK
84385-500ML 84385-6X500ML 84385-1L 84385-2.5L
Salpetersyra ≥69,0%, TraceSELECT™ för spåranalys, Fluka™
Salpetersyra
TraceSELECT™ Ultra and TraceSELECT
  • Ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • Sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • The acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions
Formel: HNO₃
MW: 63.01 g/mol
Kokpunkt: 122 °C (1.013 hPa)
Smältpunkt: –38 °C
Densitet: 1.420 g/cm3 (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011349
CAS nummer: 7697-37-2
UN: 2031
ADR: 8 II
Merck Index: 13,06608
Order Now
Specifikation testresultat
Assay 67 - 70 %
Silver (Ag) Max. 0.1 ppb
Aluminium (Al) Max. 1 ppb
Arsenic (As) Max. 0.5 ppb
Gold (Au) Max. 0.1 ppb
Boron (B) Max. 1 ppb
Barium (Ba) Max. 0.1 ppb
Beryllium (Be) Max. 0.1 ppb
Bismuth (Bi) Max. 0.1 ppb
Calcium (Ca) Max. 1 ppb
Cadmium (Cd) Max. 0.5 ppb
Cerium (Ce) Max. 0.1 ppb
Cobalt (Co) Max. 0.5 ppb
Chromium (Cr) Max. 1 ppb
Cesium (Cs) Max. 0.1 ppb
Copper (Cu) Max. 0.5 ppb
Dysprosium (Dy) Max. 0.1 ppb
Erbium (Er) Max. 0.1 ppb
Europium (Eu) Max. 0.1 ppb
Iron (Fe) Max. 1 ppb
Gallium (Ga) Max. 0.1 ppb
Gadolinium (Gd) Max. 0.1 ppb
Germanium (Ge) Max. 0.1 ppb
Hafnium (Hf) Max. 0.1 ppb
Mercury (Hg) Max. 0.1 ppb
Holmium (Ho) Max. 0.1 ppb
Indium (In) Max. 0.1 ppb
Potassium (K) Max. 1 ppb
Lanthanum (La) Max. 0.1 ppb
Lithium (Li) Max. 0.1 ppb
Lutetium (Lu) Max. 0.1 ppb
Magnesium (Mg) Max. 1 ppb
Manganese (Mn) Max. 0.1 ppb
Molybdenum (Mo) Max. 0.1 ppb
Sodium (Na) Max. 1 ppb
Niobium (nb) Max. 0.1 ppb
Neodymium (nd) Max. 0.1 ppb
Nickel (Ni) Max. 0.5 ppb
Lead (Pb) Max. 0.1 ppb
Palladium (Pd) Max. 0.5 ppb
Praseodymium (Pr) Max. 0.1 ppb
Platinum (Pt) Max. 0.5 ppb
Rubidium (Rb) Max. 0.1 ppb
Rhenium (Re) Max. 0.1 ppb
Rhodium (Rh) Max. 0.5 ppb
Ruthenium (Ru) Max. 0.5 ppb
Antimony (Sb) Max. 0.5 ppb
Scandium (Sc) Max. 0.1 ppb
Selenium (Se) Max. 1 ppb
Samarium (Sm) Max. 0.1 ppb
Tin (Sn) Max. 0.5 ppb
Strontium (Sr) Max. 0.1 ppb
Tantalum (Ta) effective ppb
Terbium (Tb) Max. 0.1 ppb
Tellurium (Te) Max. 0.1 ppb
Thorium (Th) Max. 0.1 ppb
Titanium (Ti) Max. 0.5 ppb
Thallium (Tl) Max. 0.1 ppb
Thulium (Tm) Max. 0.1 ppb
Uran (U) Max. 0.1 ppb
Vanadium (V) Max. 0.5 ppb
Tungsten (W) Max. 0.1 ppb
Yttrium (Y) Max. 0.1 ppb
Ytterbium (Yb) Max. 0.1 ppb
Zinc (Zn) Max. 0.5 ppb
Zirconium (Zr) Max. 0.1 ppb
Chloride (Cl) Max. 0.2 ppm
Phosphate (PO₄) Max. 0.01 ppm
Sulfate (SO₄) Max. 0.3 ppm
APHA Max. 10