3,3 ', 5,5'-tetrametylbensidin lösning, standard sensitivity ready-to-use
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
J63202.XE J63202.AP J63202.AU
J63202.XEEA 4100 SEK
J63202.XE J63202.AP J63202.AU
3,3 ', 5,5'-tetrametylbensidin lösning, standard sensitivity ready-to-use
3,3 ', 5,5'-tetrametylbensidin
Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result.
Formel: C₁₆H₂₀N₂
MW: 240.35 g/mol
Kokpunkt: 402
Storage Temperature: Kylskåp
MDL Number: MFCD00007748
CAS nummer: 54827-17-7
ADR: 3 II