Tin plating, pulver, electroless part A
Leverantör: Thermo Fisher Scientific
Fara
44176.TD 44176.XA
44176.TDEA 3150 SEK
44176.TD 44176.XA
Tin plating, pulver, electroless part A
Tin plating
Used to preserve solderability of printed circuit boards, for plating electronic parts, and for corrosion protection.
Storage Temperature: Omgivande
MDL Number: MFCD03703330
UN: 1759
ADR: 8,III