D (+)-Maltos monohydrat 92+% Torr_basis, ultrapure
Leverantör: Thermo Fisher Scientific
Synonyms: Maltobiose hydrate, Malt Sugar, beta-Maltose monohydrate, Maltose monohydrate, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, 4-O-α-D-Glucopyranosyl-D-glucose hydrate
J18725.A1 J18725.A7
J18725.A1EA 2150 SEK
J18725.A1 J18725.A7
D (+)-Maltos monohydrat 92+% Torr_basis, ultrapure
D (+)-Maltos monohydrat
Formel: C₁₂H₂₂O₁₁·1H₂O
MW: 360.32 g/mol
Smältpunkt: ∼ 120 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00149343
CAS nummer: 6363-53-7
EINECS: 200-716-5
Merck Index: 14,05714
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f861d7adf53134a0c485b5942d3be34c4d0b571&vid=16e5c4425a7a8e158a4200470a45483c7ff5781e&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Assay Glucose Content (Dry Basis) ≤3.0%
Assay Maltose Content (Dry Basis) ≥92.0%
Heavy Metals (Pb) ≤5 ppm