4-[(tert-Butoxycarbonylamino)methyl]benzoic acid 97%
Leverantör: Thermo Fisher Scientific
Synonyms: 4-(((tert-Butoxycarbonyl)amino)methyl)benzoic acid, 4-[(Boc-amino)methyl]benzoic Acid, Boc-4-Amb-OH
H66390.06 H66390.14
H66390.06EA 1925 SEK
H66390.06 H66390.14
4-[(tert-Butoxycarbonylamino)methyl]benzoic acid 97%
4-[(tert-Butoxycarbonylamino)methyl]benzoic acid
Formel: C₁₃H₁₇NO₄
MW: 251.28 g/mol
Smältpunkt: 164…168 °C
Storage Temperature: Omgivande
MDL Number: MFCD00228182
CAS nummer: 33233-67-9