5-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate hydrate 98%
Leverantör: Thermo Scientific
Synonyms: N-Fmoc-L-glutamic acid 5-tert-butyl ester, Fmoc-Glu(OtBu)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamic Acid 5-tert-Butyl Ester, 5-tert-Butyl N-Fmoc-L-glutamate
H59169.06 H59169.14
H59169.06EA 1825 SEK
H59169.06 H59169.14
5-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate hydrate 98%
5-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate hydrate
Formel: C₂₄H₂₇NO₆
MW: 425,48 g/mol
Smältpunkt: 83…90 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00037135
CAS nummer: 71989-18-9
EINECS: 276-253-8
Order Now
Specifikation testresultat
Assay (HPLC) Min. 97.5%