ESD safety shoes, sandals, Velcro® fastening, Biomex Dynamics, Impulse Green Easy
Leverantör: ELTEN
712551/46 712551/47 712551/40 712551/41 712551/36 712551/37 712551/42 712551/43 712551/44 712551/45 712551/38 712551/39
ELTN712551/46EA 1025 SEK
ELTN712551/46 ELTN712551/47 ELTN712551/40 ELTN712551/41 ELTN712551/36 ELTN712551/37 ELTN712551/42 ELTN712551/43 ELTN712551/44 ELTN712551/45 ELTN712551/38 ELTN712551/39
ESD safety shoes, sandals, Velcro® fastening, Biomex Dynamics, Impulse Green Easy
Skor och kängor
Metal- and leather-free, lightweight ESD safety sandals made from perforated microfibre and reflective materials, skin friendly textile lining, a full padded bellows tongue and collar, plus an extremely flexible, ergonomic, S-shaped, dual-density TPU/PU sole (Biomex Dynamics).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length ESD inlay sole
  • Semi-orthopaedic, anatomically-shaped insole supports the natural heel-to-toe foot movement
  • Abrasion-proof scuff cap
  • Protective composite toecap (200 J)
  • Metal-free, puncture resistant midsole
Ideal for use in dry work areas such as industry, storage, transport, or assembly and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/green
Certifikat: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC.