Doxapram hydrochloride hydrate ≥95%
Leverantör: Cayman Chemical
Fara
Synonyms: 1-Ethyl-4-(2-morpholinoethyl)-3,3-diphenylpyrrolidin-2-one hydrochloride hydrate, 1-Ethyl-4-[2-(4-morpholinyl)ethyl]-3,3-diphenyl-2-pyrrolidinone hydrochloride hydrate, AHR 619, Doxapram HCl
23794-10 23794-25 23794-5
CAYM23794-10EA 1675 SEK
CAYM23794-10 CAYM23794-25 CAYM23794-5
Doxapram hydrochloride hydrate ≥95%
Doxapram hydrochloride hydrate
Doxapram is a central respiratory stimulant. It increases respiration and phrenic nerve activity in anesthetized cats at a dose of 1.0 mg/kg. Doxapram acts through carotid chemoreceptors, lacking activity in anesthetized cats following carotid denervation. Formulations containing doxapram have been used to increase respiration in patients with chronic pulmonary disease, respiratory depression induced by drug overdose, and to stimulate deep breathing post-anesthesia.
  • Warning - this product is not for human or veterinary use.
Formel: C₂₄H₃₀N₂O₂·HCl·H₂O
MW: 432,98 g/mol
Storage Temperature: Frys
MDL Number: MFCD09954698
CAS nummer: 7081-53-0
UN: 2811
ADR: 6.1,III