Calcein AM 90
Leverantör: Cayman Chemical
Synonyms: Calcein-AM, Calcein O,O′-diacetate tetrakis(acetoxymethyl) ester, Calcein acetoxymethyl ester, Tetrakis(acetoxymethyl) 2,2',2'',2'''-(((3',6'-diacetoxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-2',7'-diyl)bis(methylene))bis(azanetriyl))tetraacetate
14948-1 14948-500
CAYM14948-1EA 4900 SEK
CAYM14948-1 CAYM14948-500
Calcein AM 90
Calcein AM
Calcein AM is a cell-permeable dye that upon entering live cells is cleaved by intracellular esterases, leaving membrane-impermeant calcein.
  • Fluorescent indicator with absorption and emission maxima of 494 and 517 nm respectively
  • Indicator of cell viability, cell-cell communication, cytotoxicity, or changes in intracellular calcium, fluoride, iron, or mercury
  • Calcein is a substrate for multidrug resistance-associated protein 1 (MRP1) and multidrug resistance protein 3 (MDR3, P-glycoprotein 3)
Calcein AM is used to study these proteins and their regulation.
Formel: C₄₆H₄₆N₂O₂₃
MW: 994.86 g/mol
Storage Temperature: Frys
MDL Number: MFCD05861516
CAS nummer: 148504-34-1