2-Acetylthiophene 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 1-Thiophen-2-yl-ethanone, 1-(2-Thienyl)-1-ethanone, Methyl 2-thienyl ketone
102721000 102725000
ACRO102721000EA 950 SEK
ACRO102721000 ACRO102725000
2-Acetylthiophene 98%
2-Acetylthiophene
Formel: C₆H₆OS
MW: 126,18 g/mol
Kokpunkt: 214 °C (760 mmHg)
Smältpunkt: 10…11 °C
Densitet: 1,16 g/cm³
Flampunkt: 91 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005442
CAS nummer: 88-15-3
EINECS: 201-804-6
UN: 2810
ADR: 6.1,II
Order Now
Specifikation testresultat
Appearance Clear light yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5640 to 1.5660 (20°C, 589 nm)
GC ≥97.5 %