3-Bromo-4-methylthiophene 95%
Leverantör: Thermo Scientific
Varning
379960010 379960050
ACRO379960010EA 955 SEK
ACRO379960010 ACRO379960050
3-Bromo-4-methylthiophene 95%
3-Bromo-4-methylthiophene
Formel: C₅H₅BrS
MW: 177,06 g/mol
Kokpunkt: 179…183 °C (770 mmHg)
Flampunkt: 78 °C
MDL Number: MFCD00130084
CAS nummer: 30318-99-1
UN: 2810
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5770 to 1.5790 (20°C, 589 nm)
GC ≥94 %