Hexametyldisiloxan, 0.65 cSt (25°C) ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Silicone fluid, HMDSO
A11848.0F A11848.AC A11848.AE A11848.AP
A11848.0FEA 9375 SEK
A11848.0F A11848.AC A11848.AE A11848.AP
Hexametyldisiloxan, 0.65 cSt (25°C) ≥98%
Hexametyldisiloxan
Purity: 98+% CAS No.: 107-46-0 Molecular Formula: C6H18OSi2 Formula Weight: 162.38 Flash Point: -2°(28°F) Refractive Index: 1.3770 Odour: Characteristic Melting Point: -67° Boiling Point: 100-101° Beilstein Reference: 1736258 UN No.: 1993 MDL No.: MFCD00008265
Formel: C₆H₁₈OSi₂
Kokpunkt: 100…101 °C
Smältpunkt: –67 °C
Densitet: 0,764
Flampunkt: –2 °C (28 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008265
CAS nummer: 107-46-0
EINECS: 203-492-7
UN: 1993
ADR: 3,II
REACH: 01-2119496108-31
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03d0d18c44dfa536ffcc853891b6f469333a38c5&vid=4959fefab2528efb6003e1f23c57966b65984db6&prid=web&clid=SAPDAM