Pyrimidine ≥99%
Leverantör: Thermo Scientific
Varning
A15534.03 A15534.06 A15534.14
A15534.03EA 790 SEK
A15534.03 A15534.06 A15534.14
Pyrimidine ≥99%
Pyrimidine
Purity: 99% CAS No.: 289-95-2 Molecular Formula: C4H4N2 Formula Weight: 80.09 Flash Point: 31°(87°F) Refractive Index: 1.5040 Melting Point: 20-22° Boiling Point: 123-124° Beilstein Reference: 103894 Merck Reference: 14,7987 UN No.: 1993 MDL No.: MFCD00006059
Formel: C₄H₄N₂
MW: 80,09 g/mol
Kokpunkt: 123…124 °C
Smältpunkt: 20…22 °C
Densitet: 1,016
Flampunkt: 31 °C (87 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00006059
CAS nummer: 289-95-2
EINECS: 206-026-0
UN: 1993
ADR: 3,III
Merck Index: 14,7987