Pyrimidine 99%
Leverantör: Thermo Fisher Scientific
Varning
157700050 157700250
ACRO157700050EA 1500 SEK
ACRO157700050 ACRO157700250
Pyrimidine 99%
Pyrimidine
Formel: C₄H₄N₂
Kokpunkt: 123…124 °C (760 mmHg)
Smältpunkt: 20…22 °C
Densitet: 1,01 g/cm³
Flampunkt: 31 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006059
CAS nummer: 289-95-2
EINECS: 206-026-0
UN: 1993
ADR: 3,III
Merck Index: 12,08170
Order Now
Specifikation testresultat
Appearance Clear colorless to orange Liquid or solid
Infrared spectrum Conforms
Refractive index 1.5010 to 1.5050 (20°C, 589 nm)
GC ≥98.5 %
Phosphate (PO4) ≤0.0005 %
Sulfate (SO4) ≤0.005 %
Ammonium (NH4) ≤0.003 %
Barium (Ba) ≤0.005 %
Calcium (Ca) ≤0.01 %
Iron (Fe) ≤0.0005 %
Manganese (Mn) ≤0.0005 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Strontium (Sr) ≤0.005 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %