2,4-Diamino-6-hydroxypyrimidine 96%
Leverantör: Thermo Scientific
Varning
Synonyms: 6-Aminoisocytosin, 2,4-Diamino-6-pyrimidinol, 2,4-Diaminopyrimidin-6-ol
112241000 112240250
ACRO112241000EA 1250 SEK
ACRO112241000 ACRO112240250
2,4-Diamino-6-hydroxypyrimidine 96%
2,4-Diamino-6-hydroxypyrimidine
Formel: C₄H₆N₄O
MW: 126,12 g/mol
Kokpunkt: 373 °C (1013 hPa)
Smältpunkt: 270…290 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00006098
CAS nummer: 56-06-4
EINECS: 200-254-4
Merck Index: 13,03001
Order Now
Specifikation testresultat
Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Titration with HClO4 ≥98.5 %