2-Aminopyrimidine 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Pyrimidin-2-ylamine, Pyrimidin-2-amine
104580050 104581000 104585000
ACRO104580050EA 420 SEK
ACRO104580050 ACRO104581000 ACRO104585000
2-Aminopyrimidine 98%
2-Aminopyrimidine
Formel: C₄H₅N₃
MW: 95,1 g/mol
Kokpunkt: 158 °C (186mbar)
Smältpunkt: 124…127 °C
Flampunkt: 103 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006089
CAS nummer: 109-12-6
EINECS: 203-648-4
Order Now
Specifikation testresultat
Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 123 to 127 °C
Titration with HClO4 ≥97.5 %
BSC Certification Passes