2-Bromopyrimidine ≥98%
Leverantör: Thermo Fisher Scientific
Varning
A13791.14 A13791.03 A13791.06 A13791.22
A13791.14EA 5550 SEK
A13791.14 A13791.03 A13791.06 A13791.22
2-Bromopyrimidine ≥98%
2-Bromopyrimidine
Purity: 98+% CAS No.: 4595-60-2 Molecular Formula: C4H3BrN2 Formula Weight: 158.99 Storage Temperature: 2°C to 8°C Melting Point: 53-57° Boiling Point: 62-64°/1.5mm Beilstein Reference: 605945 MDL No.: MFCD00014601
Formel: C₄H₃BrN₂
Kokpunkt: 62…64 °C (1,5 mmHg)
Smältpunkt: 53…57 °C
Densitet: >1 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00014601
CAS nummer: 4595-60-2
EINECS: 224-993-7