2-Amino-4-methylpyridine 98%
Leverantör: Thermo Fisher Scientific
Fara
A14177.14 A14177.22 A14177.36
A14177.14EA 595 SEK
A14177.14 A14177.22 A14177.36
2-Amino-4-methylpyridine 98%
2-Amino-4-methylpyridine
Purity: 98% CAS No.: 695-34-1 Molecular Formula: C6H8N2 Formula Weight: 108.14 Flash Point: 118°(244°F) Odour: Unpleasant Boiling Point: 231° Beilstein Reference: 107066 Merck Reference: 14,466 UN No.: 2811 MDL No.: MFCD00006321
Formel: C₆H₈N₂
Kokpunkt: 231 °C
Smältpunkt: 97…101 °C
Flampunkt: 118 °C (244 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00006321
CAS nummer: 695-34-1
EINECS: 211-780-9
UN: 2811
ADR: 6.1,III
Merck Index: 14,466