3-Acetylpyridine 98%
Leverantör: Thermo Scientific
Fara
Synonyms: Methyl 3-pyridyl ketone, 1-(Pyridin-3-yl)ethanone
102690100 102691000 102695000
ACRO102690100EA 535 SEK
ACRO102690100 ACRO102691000 ACRO102695000
3-Acetylpyridine 98%
3-Acetylpyridine
Formel: C₇H₇NO
MW: 121,14 g/mol
Kokpunkt: 220 °C (760 mmHg)
Smältpunkt: 12…13 °C
Densitet: 1,1 g/cm³
Flampunkt: 104 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00006396
CAS nummer: 350-03-8
EINECS: 206-496-7
UN: 2810
ADR: 6.1,III
Merck Index: 12,06195
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5320 to 1.5350 (20°C, 589 nm)
GC ≥97.5 %