2-Acetylpyridine 98%
Leverantör: Thermo Fisher Scientific
Varning
102680250 102681000 102685000
ACRO102680250EA 865 SEK
ACRO102680250 ACRO102681000 ACRO102685000
2-Acetylpyridine 98%
2-Acetylpyridine
Formel: C₇H₇NO
Kokpunkt: 188…189 °C (760 mmHg)
Smältpunkt: 8…10 °C
Densitet: 1,08 g/cm³
Flampunkt: 76 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006303
CAS nummer: 1122-62-9
EINECS: 214-355-6
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5200 to 1.5250 (20°C, 589 nm)
GC ≥97.5 %