2-Klor-6-metylpyridin 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 6-Chloro-2-picoline
152230100 152230010
ACRO152230100EA 1025 SEK
ACRO152230100 ACRO152230010
2-Klor-6-metylpyridin 98%
2-Klor-6-metylpyridin
Formel: C₆H₆ClN
MW: 127.57 g/mol
Kokpunkt: 64…68 °C (10 mmHg)
Densitet: 1.16 g/cm³
Flampunkt: 73 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006245
CAS nummer: 18368-63-3
EINECS: 242-241-6
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow to light orange Liquid
Infrared spectrum Conforms
Refractive index 1.5250 to 1.5290 (20°C, 589 nm)
GC ≥97.5 %