4-Pyridinethiol 96%
Leverantör: Thermo Scientific
Varning
Synonyms: Pyridine-4-thiol, 4-Mercaptopyridine
164710100 164710010
ACRO164710100EA 3375 SEK
ACRO164710100 ACRO164710010
4-Pyridinethiol 96%
4-Pyridinethiol
Formel: C₅H₅NS
MW: 111,17 g/mol
Smältpunkt: 182…189 °C
Storage Temperature: Frys
MDL Number: MFCD00006422
CAS nummer: 4556-23-4
EINECS: 224-926-1
Order Now
Specifikation testresultat
Appearance Yellow to yellow-green to brown Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥95 %