4-(Dimethylamino)pyridine ≥99.0%

Leverantör: TCI
Fara

Synonyms: 4-Dimethylaminopyridine, DMAP, N,N-Dimethylpyridin-4-amine

D1450-100G D1450-25G D1450-500G
TCIAD1450-100GEA 990 SEK
TCIAD1450-100G TCIAD1450-25G TCIAD1450-500G
4-(Dimethylamino)pyridine ≥99.0%
4-(Dimethylamino)pyridine
Formel: C₇H₁₀N₂
MW: 122,17 g/mol
Kokpunkt: 190°C
Smältpunkt: 109…111 °C
Flampunkt: 124 °C
MDL Number: MFCD00006418
CAS nummer: 1122-58-3
EINECS: 214-353-5
UN: 2811
ADR: 6.1,I
Merck Index: 14,03389
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ca7ad9c7c1a19b5b83e5a404f61ac9d5c914575&vid=8ca7ad9c7c1a19b5b83e5a404f61ac9d5c914575&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to lump
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 110.0 to 114.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR