2,4,6-Trifluoropyridine ≥97%
Leverantör: Thermo Scientific
|
Fara
|
L19541.03
L19541.14
L19541.03EA
975
SEK
L19541.03
L19541.14
2,4,6-Trifluoropyridine ≥97%
2,4,6-Trifluoropyridine
Purity: 97%
CAS No.: 3512-17-2
Molecular Formula: C5H2F3N
Formula Weight: 133.07
Flash Point: 17°(62°F)
Refractive Index: 1.4130
Boiling Point: 102°
Beilstein Reference: 1364338
UN No.: 2924
MDL No.: MFCD03001161
Formel:
C₅H₂F₃N Kokpunkt: 102 °C Densitet: 1,382 Flampunkt: 17 °C (62 °F) Storage Temperature: Omgivande |
MDL Number:
MFCD03001161 CAS nummer: 3512-17-2 UN: 2924 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...