Pyridine-2,3-dicarboxylic anhydride ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2,3-Pyridinedicarboxylicanhydride, 2,3-Pyridinedicarboxylic anhydride, Quinolinic anhydride
B25118.06 B25118.14 B25118.22
B25118.06EA 750 SEK
B25118.06 B25118.14 B25118.22
Pyridine-2,3-dicarboxylic anhydride ≥98%
Pyridine-2,3-dicarboxylic anhydride
Purity: 98% CAS No.: 699-98-9 Molecular Formula: C7H3NO3 Formula Weight: 149.11 Melting Point: 134-138° Beilstein Reference: 125111
Formel: C₇H₃NO₃
Kokpunkt: 360 °C (1013 hPa)
Smältpunkt: 134…138 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005915
CAS nummer: 699-98-9
EINECS: 211-834-1