Pyridine-3,5-dicarboxylic acid 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Dinicotinic acid, 3,5-Pyridinedicarboxylic acid
131900100 131900500
ACRO131900100EA 1575 SEK
ACRO131900100 ACRO131900500
Pyridine-3,5-dicarboxylic acid 98%
Pyridine-3,5-dicarboxylic acid
Formel: C₇H₅NO₄
MW: 167,12 g/mol
Kokpunkt: 357 °C (1013 hPa)
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006393
CAS nummer: 499-81-0
EINECS: 207-893-8
UN: 2811
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
GC ≥96.0 %