Pyridine-2,3-dicarboxylic acid ≥99%
Leverantör: Thermo Fisher Scientific
Varning
A11414.14 A11414.22
A11414.14EA 725 SEK
A11414.14 A11414.22
Pyridine-2,3-dicarboxylic acid ≥99%
Pyridine-2,3-dicarboxylic acid
Purity: 99% CAS No.: 89-00-9 Molecular Formula: C7H5NO4 Formula Weight: 167.12 Melting Point: ca 188° dec. Beilstein Reference: 137110 Merck Reference: 14,8073 MDL No.: MFCD00006295
Formel: C₇H₅NO₄
Kokpunkt: 357 °C (1013 hPa)
Smältpunkt: ∼ 188 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00006295
CAS nummer: 89-00-9
EINECS: 201-874-8
Merck Index: 14,8073