2-Aminonicotinic acid 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Aminopyridine-3-carboxylic acid, 2-Amino-3-pyridinecarboxylic acid
104160050 104160250
ACRO104160050EA 715 SEK
ACRO104160050 ACRO104160250
2-Aminonicotinic acid 98%
2-Aminonicotinic acid
Formel: C₆H₆N₂O₂
MW: 138,13 g/mol
Smältpunkt: 295…297 °C
Densitet: 1,545 g/cm³ (20 °C)
MDL Number: MFCD00006318
CAS nummer: 5345-47-1
EINECS: 226-296-3
Order Now
Specifikation testresultat
Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
GC ≥96.0 %