Isonicotinic acid 99%
Leverantör: Thermo Fisher Scientific
Varning
122591000 122595000 122590025
ACRO122591000EA 740 SEK
ACRO122591000 ACRO122595000 ACRO122590025
Isonicotinic acid 99%
Isonicotinic acid
Formel: C₆H₅NO₂
Kokpunkt: 260 °C (15 mmHg)
Smältpunkt: 310…315 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006429
CAS nummer: 55-22-1
EINECS: 200-228-2
Merck Index: 13,05206
Order Now
Specifikation testresultat
Appearance White to cream Powder
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %
HPLC ≥97.5 %
Specific optical rotation -190° to -204° (20°C, 589 nm) (c=2,acetone)